  • Search Results
  • Current landscape, unmet needs, and future directions for treatment of bipolar depression.
Current landscape, unmet needs, and future directions for treatment of bipolar depression.
Journal of affective disorders (2014-12-24)
Mark A Frye, Miguel L Prieto, William V Bobo, Simon Kung, Marin Veldic, Renato D Alarcon, Katherine M Moore, Doo-Sup Choi, Joanna M Biernacka, Susannah J Tye
ABSTRACT
Depression is the predominant pole of illness disability in bipolar disorder and, compared with acute mania, has less systematic research guiding treatment development. The aim of this review is to present the therapeutic options currently available for managing bipolar depression and to highlight areas of unmet need and future research. Literature search of PubMed, PsycINFO, and Cochrane databases and bibliographies from 2000 to August 2013 for treatments that have regulatory approval for bipolar depression or early controlled preliminary data on efficacy. Treatment options for bipolar depression have increased over the last decade, most notably with regulatory approval for olanzapine/fluoxetine combination, quetiapine, and lurasidone. Conventional mood stabilizers lamotrigine and divalproex have meta-analyses suggesting acute antidepressant response. Manual-based psychotherapies also appear to be effective in treating bipolar depression. The therapeutic utility of unimodal antidepressants, as a class, for the treatment of patients with bipolar depression, as a group, remains to be confirmed. There is a substantially unmet need to develop new interventions that are efficacious, effective, and have low side effect burden. Additional compounds are currently being developed that may ultimately be applicable to the treatment of bipolar depression and early open-trial data encourage further studies, but both of these topics are beyond the scope of this review. Future registrational trials will need to establish initial efficacy, but increasing interest for personalized or individualized medicine will encourage further studies on individual predictors or biomarkers of response.
Sigma-Aldrich
Lamotrigine, ≥98%, powder
Lamotrigine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Lamotrigine, United States Pharmacopeia (USP) Reference Standard
Lamotrigine for peak identification, European Pharmacopoeia (EP) Reference Standard
Lamotrigine, European Pharmacopoeia (EP) Reference Standard
Supelco
Lamotrigine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Fluoxetine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Lamotrigine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®