  • Search Results
  • Induced phenotype targeted therapy: radiation-induced apoptosis-targeted chemotherapy.
Induced phenotype targeted therapy: radiation-induced apoptosis-targeted chemotherapy.
Journal of the National Cancer Institute (2014-12-17)
Beom Suk Lee, Yong Woo Cho, Gui Chul Kim, Do Hee Lee, Chang Jin Kim, Hee Seup Kil, Dae Yoon Chi, Youngro Byun, Soon Hong Yuk, Kwangmeyung Kim, In-San Kim, Ick Chan Kwon, Sang Yoon Kim
ABSTRACT
Tumor heterogeneity and evolutionary complexity may underlie treatment failure in spite of the development of many targeted agents. We suggest a novel strategy termed induced phenotype targeted therapy (IPTT) to simplify complicated targets because of tumor heterogeneity and overcome tumor evolutionary complexity. We designed a caspase-3 specific activatable prodrug, DEVD-S-DOX, containing doxorubicin linked to a peptide moiety (DEVD) cleavable by caspase-3 upon apoptosis. To induce apoptosis locally in the tumor, we used a gamma knife, which can irradiate a very small, defined target area. The in vivo antitumor activity of the caspase-3-specific activatable prodrug combined with radiation was investigated in C3H/HeN tumor-bearing mice (n = 5 per group) and analyzed with the Student's t test or Mann-Whitney U test. All statistical tests were two-sided. We confirmed the basic principle using a caspase-sensitive nanoprobe (Apo-NP). A single exposure of radiation was able to induce apoptosis in a small, defined region of the tumor, resulting in expression of caspase-3. Caspase-3 cleaved DEVD and activated the prodrug. The released free DOX further activated DEVD-S-DOX by exerting cytotoxic effects on neighboring tumor or supporting cells, which repetitively induced the expression of caspase-3 and the activation of DEVD-S-DOX. This sequential and repetitive process propagated the induction of apoptosis. This novel therapeutic strategy showed not only high efficacy in inhibiting tumor growth (14-day tumor volume [mm(3)] vs radiation alone: 848.21 ± 143.24 vs 2511.50 ± 441.89, P < .01) but also low toxicity to normal cells and tissues. Such a phenotype induction strategy represents a conceptually novel approach to overcome tumor heterogeneity and complexity as well as to substantially improve current conventional chemoradiotherapy with fewer sequelae and side effects.
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Endoproteinase Arg-C from mouse submaxillary gland, suitable for protein sequencing, lyophilized powder