MilliporeSigma
  • Water used to moisten vegetables is a source of Escherichia coli and protozoan parasite contamination at markets in Hanoi, Vietnam.
Water used to moisten vegetables is a source of Escherichia coli and protozoan parasite contamination at markets in Hanoi, Vietnam.
Journal of water and health (2014-12-05)
Nguyen Thuy Tram, Anders Dalsgaard
ABSTRACT
The study was done to assess the level of fecal (Escherichia coli) and protozoan parasite (Cryptosporidium spp. and Giardia spp.) contamination in water used by traders to moisten vegetables at markets in Hanoi, Vietnam. A total of 200 splashing water samples from markets located within eight districts were analyzed for E. coli and Cryptosporidium spp. and Giardia spp. (oo)cysts. Giardia cysts were found in 17 splashing water samples and Cryptosporidium oocysts in nine samples, with median values of 20 cysts ml(-1) and 10 oocysts ml(-1), respectively. E. coli was found with a median concentration of 636 cfu ml(-1) and its occurrence was negatively correlated with the numbers of protozoan parasites. The splashing water was kept in buckets that were rarely cleaned and often used for handwashing. The finding of these pathogens in splashing water is likely to represent real food safety hazards.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Supelco
Water, ACS reagent, for ultratrace analysis
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized