  • Search Results
  • Phenylbutyrate induces cathelicidin expression via the vitamin D receptor: Linkage to inflammatory and growth factor cytokines pathways.
Phenylbutyrate induces cathelicidin expression via the vitamin D receptor: Linkage to inflammatory and growth factor cytokines pathways.
Molecular immunology (2014-12-03)
Nikhil N Kulkarni, Zhiqian Yi, Carolin Huehnken, Birgitta Agerberth, Gudmundur H Gudmundsson
ABSTRACT
Antimicrobial peptides (AMPs) constitute an indispensable arm of innate immunity against infectious microbes in humans. Induction of endogenous AMPs may become an alternative therapy against infections. Our previous studies have demonstrated phenylbutyrate (PBA) as a novel inducer of the AMPs cathelicidin (encoded by the CAMP gene) and human beta-defensin-1 in the human bronchial epithelial cell line VA10. In this work, we have continued by studying molecular mechanisms of PBA mediated induction of LL-37 expression and associated pathways in the human bronchial epithelial cell line VA10. In this study we demonstrate vitamin D receptor (VDR) as a key transcription factor required for PBA mediated up-regulation of the CAMP gene expression. PBA also increases mRNA expression of the vitamin D3 regulated genes CYP24A1 and CD14. The siRNA knockdown of VDR reduced PBA mediated increase in CAMP, CYP24A1 and CD14 expression. Furthermore, we demonstrate that PBA enhances Toll-Like Receptor 5 ligand flagellin regulated mRNA expression of the inflammatory cytokine TNFα and chemokine CXCL8. PBA also up-regulates the expression of the genes encoding the growth factor cytokines transforming growth factor (TGF) α, TGFβ1 and TGFβ2. Our results indicate that TGFβ type I receptor and epidermal growth factor receptor are involved in PBA mediated CAMP regulation. Finally, we show that co-treatment with PBA and vitamin D3 reduces Pseudomonas aeruginosa growth in vitro.
Product Description
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Triciribine hydrate, ≥97% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Itraconazole, ≥98% (TLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Itraconazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sodium phenylbutyrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
4-Phenylbutyric acid, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Cholecalciferol (D3), analytical standard
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)