  • Search Results
  • Metal levels in eggs of waterbirds in the New York Harbor (USA): trophic relationships and possible risk to human consumers.
Metal levels in eggs of waterbirds in the New York Harbor (USA): trophic relationships and possible risk to human consumers.
Journal of toxicology and environmental health. Part A (2014-11-27)
Joanna Burger, Susan Elbin
ABSTRACT
Health professionals are interested in evaluating the risks that heavy metals pose to eco-receptors and humans. The objective of this study was to examine levels of mercury (Hg), lead (Pb), cadmium (Cd), and other contaminants in waterbirds nesting in the New York harbor in 2012 to determine (1) whether there were species and locational differences, and (2) whether consumption of eggs posed a health risk to predators or humans. For arsenic (As), Pb, Hg, and selenium (Se), species contributed more to variations in levels than location; for Cd and chromium (Cr), location was more significant. Mean metal levels differed among species for all metals, except Cd. Highest levels were As (great black-backed gulls, Larus marinus), Cr (great egret, Ardea alba), Pb (Canada goose, Branta canadensis), and Hg and Se (black-crowned night heron, Nycticorax nycticorax). There were significant locational differences only for herring gulls (Larus argentatus); significant differences were found for all metals. Levels of Hg and Pb may be sufficiently high in eggs of some species to produce adverse effects in predators that eat them. The proportion of samples above 0.3 ppm Hg (U.S. Environmental Protection Agency [EPA] freshwater criteria for freshwater fish), the contaminant of health concern, ranged from 0% (Canada goose, great egret), to 14 and 27% in gulls, to 50% (black-crowned night heron). Some herring gull, great black-backed gull, and black-crowned night heron eggs had 0.5 ppm or higher Hg. Thus, human consumption of eggs may pose a risk to fetuses and young children.
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Lead, shot, <2 mm, 99.9% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Mercury, electronic grade, 99.9999% trace metals basis
Sigma-Aldrich
Lead, powder, −100 mesh, 99.95% trace metals basis
Sigma-Aldrich
Lead, shot, 1-3 mm, 99.995% trace metals basis
Sigma-Aldrich
Mercury, ≥99.99% trace metals basis
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis
Lead, foil, light tested, 50x50mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, not light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.01mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.025mm, 99.99+%
Lead, foil, light tested, 50x50mm, thickness 0.008mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.0025mm, temporary acrylic support, 99.99+%
Lead, foil, not light tested, 100x100mm, thickness 0.025mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.05mm, as rolled, 97% (Sn 1.5%, Sb 1.5%)
Lead, foil, light tested, 25x25mm, thickness 0.008mm, 99.99+%
Lead, foil, not light tested, 150x150mm, thickness 0.05mm, as rolled, 96% (Sn 1.5%, Sb 2.5%)
Lead, foil, not light tested, 100x100mm, thickness 0.004mm, 99.99+%
Lead, foil, not light tested, 25x25mm, thickness 0.009mm, 99.99+%
Lead, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.99+%
Lead, wire reel, 0.1m, diameter 2.0mm, as drawn, 99.99+%
Lead, foil, light tested, 25x25mm, thickness 0.04mm, 99.99+%
Lead, rod, 200mm, diameter 19.0mm, 99.95%
Lead, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.99+%
Lead, foil, not light tested, 50x50mm, thickness 0.005mm, 99.99+%
Lead, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Lead, microfoil, disks, 10mm, thinness 0.1μm, specific density 103.4μg/cm2, permanent mylar 3.5μm support, 99.99+%