MilliporeSigma
  • Fungal metabolite myriocin promotes human herpes simplex virus-2 infection.
Fungal metabolite myriocin promotes human herpes simplex virus-2 infection.
Life sciences (2014-12-03)
Jingjing Wang, Xuancheng Guo, Ziying Yang, Ren-Xiang Tan, Xiaoqing Chen, Erguang Li
ABSTRACT
Myriocin is a fungal metabolite with antiviral activity, including influenza, hepatitis B, and hepatitis C viruses. We investigated whether myriocin has activity against human HSV-2, one of the most prevalent pathogens of sexually transmitted disease. Cell culture systems were used to evaluate myriocin effect on HSV-2 infection. Plaque forming assay and immunoblotting studies were used to determine virus production and viral protein expression, respectively. Myriocin showed no cytotoxic effect at up to 5 μM. Myriocin treatment did not inhibit HSV-2 infection. Instead, the treatment resulted in accelerated replication of HSV-2 and increased titers of infectious virion. The effect was detected at concentrations as low as 3 nM and plateaued at approximately 30 nM. Myriocin at 30 nM increased HSV-2 production by approximately 1.7 logs. Myriocin also promoted HSV-1 infection but required higher concentrations. A time course study revealed that myriocin promoted HSV-2 infection by acceleration of virus replication. Unlike trichostatin A that promotes HSV-2 infection and histone modifications, myriocin treatment did not alter histone modifications. Myriocin is a well characterized inhibitor of sphingolipid biosynthesis pathway. Structurally different inhibitors of the pathway showed no effect on HSV-2 infection. Exogenous sphingolipids did not reverse the effect of myriocin on HSV-2 infection either. We found that myriocin promotes HSV-2 replication at nanomolar concentrations with yet unknown mechanisms. Further studies may uncover novel mechanisms regulating HSV replication and targets of myriocin action. This may have potential application in enhancing efficacy of oncolytic HSV for cancer therapy and other diseases.
Product Description
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
D-Sphingosine, synthetic
Sigma-Aldrich
Myriocin from Mycelia sterilia, ≥98% (HPLC), powder
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
D-Sphingosine, ≥98.0% (TLC)