  • Search Results
  • Complement protein C1q bound to apoptotic cells suppresses human macrophage and dendritic cell-mediated Th17 and Th1 T cell subset proliferation.
Complement protein C1q bound to apoptotic cells suppresses human macrophage and dendritic cell-mediated Th17 and Th1 T cell subset proliferation.
Journal of leukocyte biology (2014-11-09)
Elizabeth V Clarke, Brian M Weist, Craig M Walsh, Andrea J Tenner
ABSTRACT
A complete genetic deficiency of the complement protein C1q results in SLE with nearly 100% penetrance in humans, but the molecular mechanisms responsible for this association have not yet been fully determined. C1q opsonizes ACs for enhanced ingestion by phagocytes, such as Mϕ and iDCs, avoiding the extracellular release of inflammatory DAMPs upon loss of the membrane integrity of the dying cell. We previously showed that human monocyte-derived Mϕ and DCs ingesting autologous, C1q-bound LALs (C1q-polarized Mϕ and C1q-polarized DCs), enhance the production of anti-inflammatory cytokines, and reduce proinflammatory cytokines relative to Mϕ or DC ingesting LAL alone. Here, we show that C1q-polarized Mϕ have elevated PD-L1 and PD-L2 and suppressed surface CD40, and C1q-polarized DCs have higher surface PD-L2 and less CD86 relative to Mϕ or DC ingesting LAL alone, respectively. In an MLR, C1q-polarized Mϕ reduced allogeneic and autologous Th17 and Th1 subset proliferation and demonstrated a trend toward increased Treg proliferation relative to Mϕ ingesting LAL alone. Moreover, relative to DC ingesting AC in the absence of C1q, C1q-polarized DCs decreased autologous Th17 and Th1 proliferation. These data demonstrate that a functional consequence of C1q-polarized Mϕ and DC is the regulation of Teff activation, thereby "sculpting" the adaptive immune system to avoid autoimmunity, while clearing dying cells. It is noteworthy that these studies identify novel target pathways for therapeutic intervention in SLE and other autoimmune diseases.
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, for molecular biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, for molecular biology, 2 M in H2O
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
5-Carboxy-fluorescein diacetate N-succinimidyl ester, for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate N-succinimidyl ester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Complement component C1q from human serum, ≥95% (SDS-PAGE)