  • Search Results
  • The molecular mechanisms of a novel multi-kinase inhibitor ZLJ33 in suppressing pancreatic cancer growth.
The molecular mechanisms of a novel multi-kinase inhibitor ZLJ33 in suppressing pancreatic cancer growth.
Cancer letters (2014-10-11)
Yan Li, Ke Tang, Lijing Zhang, Chao Li, Fei Niu, Wanqi Zhou, Hanze Yang, Zhiqiang Feng, Xiaoguang Chen
ABSTRACT
ZLJ33, an oral active multi-kinase inhibitor, was evaluated both in vitro and in vivo against human pancreatic cancer. It could effectively inhibit cell proliferation, induce apoptosis, and cause inhibition of invasion in pancreatic cancer cells. At a dose of 15.0 mg/kg, ZLJ33 induced tumor shrink in Mia-PaCa2, Capan2, and AsPC-1 xenografts models by 60.59%, 74.19%, and 71.54% according to the tumor weight, respectively. The effect of ZLJ33 on pancreatic cancer was mainly mediated by inactivation of p-PDGFRβ, p-c-Raf, and p-RET. Treatment with ZLJ33 did not cause side effect of hematology indexes in the pancreatic cancer xenograft model. ZLJ33 could be a potential therapeutic agent against pancreatic cancer.
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Supelco
Nicotinamide (Niacinamide), analytical standard