  • Search Results
  • Treatment response to antioxidants and zinc based on CFH and ARMS2 genetic risk allele number in the Age-Related Eye Disease Study.
Treatment response to antioxidants and zinc based on CFH and ARMS2 genetic risk allele number in the Age-Related Eye Disease Study.
Ophthalmology (2014-09-10)
Carl C Awh, Steven Hawken, Brent W Zanke
ABSTRACT
To evaluate the impact of complement factor H (CFH) and age-related maculopathy susceptibility 2 (ARMS2) risk alleles on the observed response to components of the Age-Related Eye Disease Study (AREDS) formulation. Genetic and statistical subgroup analysis of a randomized, prospective clinical trial. White patients from the AREDS with category 3 or 4 age-related macular degeneration (AMD) with available DNA (n = 989). Four genotype groups based on CFH and ARMS2 risk allele number were defined. Progression to advanced AMD was analyzed by genotype and treatment using Cox proportionate hazards estimates and 7-year events. The effect of predefined genotype group on treatment-specific progression to advanced AMD. Patients with 2 CFH risk alleles and no ARMS2 risk alleles progressed more with zinc-containing treatment compared with placebo, with a hazard ratio (HR) of 3.07 (P = 0.0196) for zinc and 2.73 (P = 0.0418) for AREDS formulation (AF). Seven-year treatment-specific progression rates were: placebo, 17.0%; zinc, 43.2% (P = 0.023); and AF, 40.2% (P = 0.039). Patients with 0 or 1 CFH risk alleles and 1 or 2 ARMS2 risk alleles benefited from zinc-containing treatment compared with placebo, with an HR of 0.514 for zinc (P = 0.012) and 0.569 for AF (P = 0.0254). Seven-year treatment-specific AMD progression rates were as follows: placebo, 43.3%; zinc, 25.2% (P = 0.020); and AF, 27.3% (P = 0.011). Zinc and AF treatment each interacted statistically with these 2 genotype groups under a Cox model, with P values of 0.000999 and 0.00366, respectively. For patients with 0 or 1 CFH risk alleles and no ARMS2 risk alleles, neither zinc-containing treatment altered progression compared with placebo, but treatment with antioxidants decreased progression (HR, 0.380; P = 0.034). Seven-year progression with placebo was 22.6% and with antioxidants was 9.17% (P = 0.033). For patients with 2 CFH risk alleles and 1 or 2 ARMS2 risk alleles, no treatment was better than placebo (48.4%). The benefit of the AREDS formulation seems the result of a favorable response by patients in only 1 genotype group, balanced by neutral or unfavorable responses in 3 genotype groups.
Product Description
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Sodium ascorbate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
β-Carotene, synthetic, ≥93% (UV), powder
Sigma-Aldrich
β-Carotene, synthetic, ≥95% (HPLC), crystalline
Supelco
β-Carotene, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(+)-Sodium L-ascorbate, analytical standard
Supelco
L-Ascorbic acid, analytical standard
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Complement factor H from human plasma, 1 mg/mL in PBS, pH 7.2, ≥90% (SDS-PAGE)
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material