  • Search Results
  • Assessment of platelet function with light transmission aggregometry in 24 patients supported with a continuous-flow left ventricular assist device: a single-center experience.
Assessment of platelet function with light transmission aggregometry in 24 patients supported with a continuous-flow left ventricular assist device: a single-center experience.
The Journal of thoracic and cardiovascular surgery (2014-09-13)
Mathieu Fiore, Chloé James, Christine Mouton, Joachim Calderon, Laurent Barandon, Alexandre Ouattara, François Picard
ABSTRACT
This study evaluated platelet function for an extended period of time in patients with a HeartMate II continuous-flow left ventricular assist device (Thoratec Corporation, Pleasanton, Calif) with light transmission aggregometry and investigated the potential role of this test in clinical management. Twenty-four patients were studied prospectively after implantation. Mean duration of support was 8.5 months. Platelet functions were assessed with light transmission aggregometry induced by thrombin receptor agonist peptide, ristocetin, or arachidonic acid. All patients received an aspirin regimen that was progressively increased until arachidonic acid-triggered platelet aggregation dropped lower than 20%. Plasma levels of von Willebrand factor were also determined when ristocetin-induced platelet agglutination was impaired. Intensity of platelet aggregation with thrombin receptor agonist peptide was little changed in patients with a HeartMate II relative to control subjects. Aspirin dose greater than 160 mg/d was progressively required in 46% of patients. Ristocetin-induced platelet agglutination was impaired in 4 patients in association with a lack of high molecular weight von Willebrand factor multimers. Three patients had thromboembolic events (12.5%) and 8 (33%) suffered from major bleeding complications. High platelet reactivity during treatment with aspirin is common in patients with a HeartMate II. Moreover, light transmission aggregometry may detect impaired ristocetin-induced platelet agglutination, enabling dosage of aspirin to be adjusted. Our strategy showed no major improvements in terms of thrombosis rate when compared with published data, although bleeding frequency was somewhat reduced. Benefits of light transmission aggregometry testing need to be assessed in a larger randomized study with a longer follow-up.
Product Description
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Arachidonic acid, >95.0% (GC)
Sigma-Aldrich
Arachidonic acid, from non-animal source, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Arachidonic acid sodium salt, from Porcine liver, ≥99% (capillary GC), waxy solid
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard