MilliporeSigma
  • Extracellular magnesium and calcium reduce myotonia in isolated ClC-1 chloride channel-inhibited human muscle.
Extracellular magnesium and calcium reduce myotonia in isolated ClC-1 chloride channel-inhibited human muscle.
Muscle & nerve (2014-04-09)
Martin Skov, Frank Vincenzo De Paoli, Jesper Lausten, Ole Baekgaard Nielsen, Thomas Holm Pedersen
ABSTRACT
Experimental myotonia induced in rat muscle by ClC-1 chloride channel-inhibited has been shown to be related inversely to extracellular concentrations of Mg(2+) and Ca(2+) ([Mg(2+) ]o and [Ca(2+) ]o) within physiological ranges. Because this implicates a role for [Mg(2+)]o and [Ca(2+)]o in the variability of symptoms among myotonia congenita patients, we searched for similar effects of [Mg(2+)]o and [Ca(2+)]o on myotonia in human muscle. Bundles of muscle fibers were isolated from abdominal rectus in patients undergoing abdominal surgery. Myotonia was induced by ClC-1 inhibition using 9-anthracene carboxylic acid (9-AC) and was assessed from integrals of force induced by 5-Hz stimulation for 2 seconds. Myotonia disappeared gradually when [Mg(2+)]o or [Ca(2+)]o were elevated throughout their physiological ranges. These effects of [Mg(2+)]o and [Ca(2+)]o were additive and interchangeable. These findings suggest that variations in symptoms in myotonia congenita patients may arise from physiological variations in serum Mg(2+) and Ca(2+).
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Magnesium, microfoil, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, 6 micron aluminum permanent support, 99.9%
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 0.5m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, wire reel, 2m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca+2 in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Magnesium, ribbon, ≥99.5% Mg basis
Sigma-Aldrich
Magnesium, ReagentPlus®, powder, −50 mesh, ≥99%
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis