  • Search Results
  • Complete oxidation of CO, ethanol, and ethyl acetate over copper oxide supported on titania and ceria modified titania.
Complete oxidation of CO, ethanol, and ethyl acetate over copper oxide supported on titania and ceria modified titania.
J. Catal. (1998)
Larsson PO and Andersson A.