  • Search Results
  • Photo-induced current and its degradation in Al4C3/Al2O3 (0001) grown by metalorganic chemical vapor deposition
Photo-induced current and its degradation in Al4C3/Al2O3 (0001) grown by metalorganic chemical vapor deposition
Thin Solid Films (2014)
Kim D, et al.