  • Search Results
  • Mercury bioaccumulation and bioaccumulation factors for Everglades mosquitofish as related to sulfate: a re-analysis of Julian II (2013).
Mercury bioaccumulation and bioaccumulation factors for Everglades mosquitofish as related to sulfate: a re-analysis of Julian II (2013).
Bulletin of environmental contamination and toxicology (2014-09-28)
Curtis D Pollman, Donald M Axelrad
ABSTRACT
The Everglades, an ecosystem of international significance, has elevated biota mercury levels representing risk to human and wildlife consumers of fish. Given the critical role of sulfate in the methylation of mercury, and because there is a significant agricultural contribution, one potential means of reducing these mercury levels is reducing Everglades sulfate inputs. Julian II (Bull Environ Contam Toxicol 90:329-332, 2013) conducted regression modeling of the relationship between surface water sulfate concentrations and Gambusia spp. mercury bioconcentration factors across the major hydrologic subunits of the Everglades, and used those results to draw conclusions about the role of sulfate in the cycling of mercury in the Everglades. We however demonstrate a number of fundamental problems with the analysis, interpretation and conclusions. As a result, we strongly caution against using the results of Julian II (Bull Environ Contam Toxicol 90:329-332, 2013) to formulate management decisions regarding mitigation of the Everglades mercury problem.
Sigma-Aldrich
Mercury, ≥99.99% trace metals basis
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, PCR Reagent
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, BioPerformance Certified
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O