  • Search Results
  • Pharmacogenetics of the DNA repair pathways in advanced non-small cell lung cancer patients treated with platinum-based chemotherapy.
Pharmacogenetics of the DNA repair pathways in advanced non-small cell lung cancer patients treated with platinum-based chemotherapy.
Cancer letters (2014-07-30)
Ivana Sullivan, Juliana Salazar, Margarita Majem, Cinta Pallarés, Elisabeth Del Río, David Páez, Montserrat Baiget, Agustí Barnadas
ABSTRACT
Genetic variants in DNA repair genes may play a role in the effectiveness of platinum-based chemotherapy in non-small cell lung cancer (NSCLC). We analyzed 17 SNPs in eight genes (ERCC1, ERCC2, ERCC3, ERCC4, ERCC5, XPA, XRCC1 and XRCC2) involved in DNA repair mechanisms and its association with outcome in NSCLC. This prospective study included patients with stages III and IV treated with platinum-based chemotherapy. All patients (n = 161) received cisplatin or carboplatin plus a third-generation drug. Additionally, stage IIIA and IIIB patients (n = 74) received concomitant or sequential radiotherapy. Germline polymorphisms were analyzed using the BioMark system in blood DNA samples. We found that in stage III patients, response was significantly associated with SNPs in ERCC1 and in ERCC3 genes, while radiotherapy-derived toxicity correlated with SNPs in the ERCC2 gene. In stage IV patients, response was associated with a genetic variant in the ERCC4 gene and survival with a SNP in the XRCC1 gene. The complexity of the DNA repair mechanisms along with the heterogeneity in the treatment of lung cancer could explain the role of multiple genes as putative biomarkers of patient outcome.
Product Description
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Carboplatin
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis