  • Search Results
  • Development of a selective agar plate for the detection of Campylobacter spp. in fresh produce.
Development of a selective agar plate for the detection of Campylobacter spp. in fresh produce.
International journal of food microbiology (2014-08-16)
Jin-Hee Yoo, Na-Young Choi, Young-Min Bae, Jung-Su Lee, Sun-Young Lee
ABSTRACT
This study was conducted to develop a selective medium for the detection of Campylobacter spp. in fresh produce. Campylobacter spp. (n=4), non-Campylobacter (showing positive results on Campylobacter selective agar) strains (n=49) isolated from fresh produce, indicator bacteria (n=13), and spoilage bacteria isolated from fresh produce (n=15) were plated on four Campylobacter selective media. Bolton agar and modified charcoal cefoperazone deoxycholate agar (mCCDA) exhibited higher sensitivity for Campylobacter spp. than did Preston agar and Hunt agar, although certain non-Campylobacter strains isolated from fresh produce by using a selective agar isolation method, were still able to grow on Bolton agar and mCCDA. To inhibit the growth of non-Campylobacter strains, Bolton agar and mCCDA were supplemented with 5 antibiotics (rifampicin, polymyxin B, sodium metabisulfite, sodium pyruvate, ferrous sulfate) and the growth of Campylobacter spp. (n=7) and non-Campylobacter strains (n=44) was evaluated. Although Bolton agar supplemented with rifampicin (BR agar) exhibited a higher selectivity for Campylobacter spp. than did mCCDA supplemented with antibiotics, certain non-Campylobacter strains were still able to grow on BR agar (18.8%). When BR agar with various concentrations of sulfamethoxazole-trimethoprim were tested with Campylobacter spp. (n=8) and non-Campylobacter (n=7), sulfamethoxazole-trimethoprim was inhibitory against 3 of 7 non-Campylobacter strains. Finally, we validated the use of BR agar containing 50mg/L sulfamethoxazole (BRS agar) or 0.5mg/L ciprofloxacin (BRCS agar) and other selective agars for the detection of Campylobacter spp. in chicken and fresh produce. All chicken samples were positive for Campylobacter spp. when tested on mCCDA, BR agar, and BRS agar. In fresh produce samples, BRS agar exhibited the highest selectivity for Campylobacter spp., demonstrating its suitability for the detection of Campylobacter spp. in fresh produce.
Sigma-Aldrich
Select agar
Millipore
Agar, for chromogenic media
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Type A, ash 5-6%
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type E, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Millipore
Agar, high purity, low ionic content, low gel strenght
Millipore
Agar, for membrane filtration, low gel strengh
Sigma-Aldrich
Bacteriological agar, for molecular biology