  • Search Results
  • Effect of lauric arginate, nisin Z, and a combination against several food-related bacteria.
Effect of lauric arginate, nisin Z, and a combination against several food-related bacteria.
International journal of food microbiology (2014-08-12)
Rinrada Pattanayaiying, Aran H-Kittikun, Catherine N Cutter
ABSTRACT
The effects of lauric arginate (LAE) and nisin Z, alone or in combination, on cell damage were investigated against Escherichia coli O157:H7, Listeria monocytogenes and Brochothrix thermosphacta, by scanning electron microscopy (SEM) and transmission electron microscopy (TEM) observations, efflux of potassium and phosphate ions, and growth inhibition. A combination of LAE with nisin Z caused severe and dramatic changes in the cytoplasmic membrane and cell lysis of both Gram-positive and Gram-negative bacteria. The combination treatment also caused significant potassium and phosphate ion leakage of E. coli O157:H7, L. monocytogenes and B. thermosphacta, when compared with other treatments: 16.62±1.05, 50.35±0.81 and 45.47±1.15mg/L of potassium ion and 122.66±8.81, 97.96±3.31 and 26.47±13.97mg/L of phosphate ion after treatment for 6h, respectively. Bacteria were reduced by approximately 7log10CFU/mL within the first hour of treatment and then cells were unable to grow for the remainder of the experiment. Treatment with LAE alone resulted in changes in cellular morphology, coagulation of the cytoplasm, and low level leakage of potassium and phosphate ions in all bacteria tested. Treatment of L. monocytogenes and B. thermosphacta with nisin Z (320AU/mL of final concentration) resulted in the formation of membrane channels and leakage of potassium and phosphate ions at rather high levels; but the bacteriocin was not effective against E. coli O157:H7. LAE or nisin Z reduced growth of both L. monocytogenes and B. thermosphacta by approximately 7log10CFU/mL. Conversely, E. coli O157:H7 was not inhibited by treatments with nisin Z, but decreased by approximately 4.45log10CFU/mL after treatment with LAE. These findings provide additional information on the mode of action of these compounds on bacterial populations.
Product Description
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Supelco
(±)-Propylene oxide, analytical standard
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
(±)-Propylene oxide, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Supelco
Ethanol standards 10% (v/v), 10 % (v/v) in H2O, analytical standard
Sigma-Aldrich
Nisin from Lactococcus lactis, potency: ≥.9 per IU/g
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
(±)-Propylene oxide, ReagentPlus®, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Supelco
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer
Supelco
Chloroform, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%