  • Search Results
  • Bacterioplanes sanyensis gen. nov., sp. nov., a PHB-accumulating bacterium isolated from a pool of Spirulina platensis cultivation.
Bacterioplanes sanyensis gen. nov., sp. nov., a PHB-accumulating bacterium isolated from a pool of Spirulina platensis cultivation.
Archives of microbiology (2014-07-21)
Guanghua Wang, Qikun Jia, Tao Li, Shikun Dai, Huanlian Wu, Hui He, Jiewei Fan, Wenzhou Xiang, Xiang Li
ABSTRACT
A Gram-negative, poly-3-hydroxybutyrate-accumulating rod bacterium, strain GYP-2(T), was isolated from a pool of marine Spirulina platensis cultivation, Sanya, China. Growth was observed at 10-45 °C and pH 6-10 in the presence of 1-10 % (w/v) NaCl. Phylogenetic analysis based on 16S rRNA gene sequences indicated that the new isolate belonged to Gammaproteobacteria and displayed 93.8-95.3 % 16S rRNA gene sequences similarities to members of the genera Thalassolituus, Oleibacter, and Oceanobacter, but house-keeping gene gyrB (encode DNA gyrase beta subunit) demonstrated that the new isolate was distantly related to Thalassolituus, Oleibacter, and Oceanobacter species (only 77-83 % gene gyrB sequences similarities).The G+C content of genomic DNA was 55 mol%. The major respiratory quinone was Q-9, while that for Oceanobacter kriegii LMG 6238(T) was Q-8. Major polar lipids were diphosphatidylglycerol, phosphatidylglycerol, and phosphatidylethanolamine. On the basis of its physiological, chemotaxonomic, and molecular properties, strain GYP-2(T) is suggested to represent a novel species of a new genus in Gammaproteobacteria, for which the name Bacterioplanes sanyensis gen. nov., sp. nov. is proposed. The type strain is GYP-2(T) (=CGMCC 1.12392(T)=KCTC 32220(T)).
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from Glycine max (soybean), Type IV, ≥98% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution
Sigma-Aldrich
3-sn-Phosphatidylethanolamine solution, from soybean, ≥97.0% (10 mg phospholipid per ml CHCl3, TLC)
Sigma-Aldrich
Poly[(R)-3-hydroxybutyric acid], natural origin
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%