  • Search Results
  • Renoprotective mechanisms of chlorogenic acid in cisplatin-induced kidney injury.
Renoprotective mechanisms of chlorogenic acid in cisplatin-induced kidney injury.
Toxicology (2014-07-22)
Robert Domitrović, Olga Cvijanović, Vesna Šušnić, Nataša Katalinić
ABSTRACT
The aim of this study was to investigate the renoprotective activity of chlorogenic acid (CA) in a murine model of cisplatin (CP)-induced kidney injury. Male BALB/cN mice were gavaged daily with CA at 3, 10 and 30mg/kg for two successive days, 48h after intraperitoneal injection of CP (13mg/kg). On the fifth day, serum creatinine and blood urea nitrogen (BUN) levels were significantly increased in CP-intoxicated mice, which was recovered by CA. Renal oxidative stress, evidenced by increased 4-hydroxynonenal (4-HNE) expression, was significantly reduced with CA. Simultaneously, the overexpression of heme oxygenase 1 (HO-1) and cytochrome P450 E1 (CYP2E1) was attenuated. The inhibition of inflammatory response by CA was achieved through the reduction of tumor necrosis factor-α (TNF-α) and cyclooxygenase-2 (COX-2) expression. Additionally, CA significantly suppressed p53, Bax active caspase-3, cyclin D1 and microtubule-associated protein 1 light chain 3 isoform B (LC3B) expression, suggesting the inhibition of both apoptosis and autophagy. The expression of multidrug resistance-associated proteins (Mrp1 and Mrp2) increased and organic cation transporter 2 (Oct2) decreased by CP, protecting the kidneys from nephrotoxicity by reducing the burden of tubular cells. CA dose-dependently restored Mrp1, Mrp2 and Oct2 expression. The recovery of kidney tissue form CP injury was accompanied by increased proliferating nuclear cell antigen (PCNA) expression. The results of this study suggest that CA attenuates CP-induced kidney injury through suppression of oxidative stress, inflammation, apoptosis and autophagy, with the improvement in kidney regeneration.
Sigma-Aldrich
Anti-CASP3 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-CASP3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-CASP3 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Anti-Caspase 3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Trometamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
SAFC
Tromethamine
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard