  • Search Results
  • FurC regulates expression of zupT for the central zinc importer ZupT of Cupriavidus metallidurans.
FurC regulates expression of zupT for the central zinc importer ZupT of Cupriavidus metallidurans.
Journal of bacteriology (2014-07-23)
Christopher Schmidt, Claudia Schwarzenberger, Cornelia Große, Dietrich H Nies
ABSTRACT
The zinc importer ZupT is required for the efficient allocation of zinc to zinc-dependent proteins in the metal-resistant bacterium Cupriavidus metallidurans but not for zinc import per se. The expression of zupT is upregulated under conditions of zinc starvation. C. metallidurans contains three members of the Fur family of regulators that qualify as candidates for the zupT regulator. The expression of a zupT-lacZ reporter gene fusion was strongly upregulated in a ΔfurC mutant but not in a ΔfurA or ΔfurB mutant. Expression of the genes for transition-metal importers (pitA, corA1, corA2, and corA3) was not changed in this pattern in all three Δfur mutants, but they were still downregulated under conditions of elevated zinc concentrations, indicating the presence of another zinc-dependent regulator. FurA was a central regulator of the iron metabolism in C. metallidurans, and furA was constitutively expressed under the conditions tested. Expression of furB was upregulated under conditions of iron starvation, and FurB could be an iron starvation Fur connecting general metal and iron homeostasis, as indicated by the phenotype of a ΔfurB ΔfurC double mutant. FurC was purified as a Strep-tagged protein and retarded the electrophoretic mobility of a DNA fragment upstream of zupT. Binding of FurC to this operator region was influenced by the presence of zinc ions and EDTA. Thus, FurC is the main zinc uptake regulator (Zur) of C. metallidurans and represses synthesis of the central zinc importer ZupT when sufficient zinc is present.
Sigma-Aldrich
Glycerol solution, puriss. p.a., 86-89% (T)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn+2 in THF, highly reactive Rieke®metal
Sigma-Aldrich
Zinc, mossy, 99.99% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, shot, 99.9999%
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Zinc, foil, 8mm disks, thickness 0.01mm, 99.9%
Zinc, foil, 50mm disks, thickness 1.0mm, as rolled, 99.99+%
Zinc, foil, 25mm disks, thickness 0.20mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.25mm, as rolled, 99.99+%
Zinc, foil, 2m coil, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 25mm disks, thickness 0.5mm, as rolled, 99.99+%
Zinc, foil, 2m coil, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, light tested, 150x150mm, thickness 0.05mm, as rolled, 99.95+%
Zinc, foil, 8mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, 6mm disks, thickness 0.175mm, as rolled, 99.95+%
Zinc, foil, not light tested, 150x150mm, thickness 0.025mm, as rolled, 99.95+%