  • Search Results
  • Habitat stress initiates changes in composition, CO2 gas exchange and C-allocation as life traits in biological soil crusts.
Habitat stress initiates changes in composition, CO2 gas exchange and C-allocation as life traits in biological soil crusts.
The ISME journal (2014-04-04)
Claudia Colesie, T G Allan Green, Ilka Haferkamp, Burkhard Büdel
ABSTRACT
Biological soil crusts (BSC) are the dominant functional vegetation unit in some of the harshest habitats in the world. We assessed BSC response to stress through changes in biotic composition, CO2 gas exchange and carbon allocation in three lichen-dominated BSC from habitats with different stress levels, two more extreme sites in Antarctica and one moderate site in Germany. Maximal net photosynthesis (NP) was identical, whereas the water content to achieve maximal NP was substantially lower in the Antarctic sites, this apparently being achieved by changes in biomass allocation. Optimal NP temperatures reflected local climate. The Antarctic BSC allocated fixed carbon (tracked using (14)CO2) mostly to the alcohol soluble pool (low-molecular weight sugars, sugar alcohols), which has an important role in desiccation and freezing resistance and antioxidant protection. In contrast, BSC at the moderate site showed greater carbon allocation into the polysaccharide pool, indicating a tendency towards growth. The results indicate that the BSC of the more stressed Antarctic sites emphasise survival rather than growth. Changes in BSC are adaptive and at multiple levels and we identify benefits and risks attached to changing life traits, as well as describing the ecophysiological mechanisms that underlie them.
Product Description
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, puriss. p.a., powder
Carbon, foil, 10mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, tube, graphite, 50mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, foil, not light tested, 150x150mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, microleaf, 25x70mm, thinness 0.50μm, specific density 100μg/cm2, temporary glass support, annealed, 99.997%
Carbon, tube, graphite, 150mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 100mm, diameter 25mm, graphite, 99.95%
Carbon, tube, graphite, 100mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 100mm, diameter 5.0mm, graphite, 100%
Carbon, rod, 150mm, diameter 3.0mm, graphite, 100%
Carbon, rod, 150mm, diameter 50mm, graphite, 99.95%
Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%
Carbon, rod, 300mm, diameter 6.35mm, graphite, 100%
Carbon, tube, graphite, 100mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, tube, graphite, 50mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 150mm, diameter 2.0mm, graphite, 100%
Carbon, foil, 5x10mm, thickness 2.0mm, hOpg
Carbon, foil, 250x250mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, microleaf, 25x70mm, thinness 1.25μm, specific density 250μg/cm2, 99.997%