  • Search Results
  • Optimization of modified Middlebrook 7H11 agar for isolation of Mycobacterium bovis from raw milk cheese.
Optimization of modified Middlebrook 7H11 agar for isolation of Mycobacterium bovis from raw milk cheese.
Letters in applied microbiology (2014-06-04)
R Forgrave, J A Donaghy, A Fisher, M T Rowe
ABSTRACT
Reports have highlighted the absence of contemporary peer reviewed publications pertaining to Mycobacterium bovis culture from raw milk and cheese. By replicating traditional methods, cheese-making methodology and equipment were devised to produce Cheddar (n = 6) and Caerphilly (n = 3) artificially contaminated with M. bovis (three genotypes) under stringent laboratory-containment guidelines for handling hazardous microbiological material. Middlebrook 7H11, modified for M. bovis isolation, was assessed for capacity to enumerate M. bovis despite changing cheese microflora and prolonged M. bovis exposure to the cheese matrix using maturing cheese test portions (n = 63; up to 16 weeks). Malachite green (MG) containing media isolated M. bovis at significantly (P < 0·05) lower levels than unmodified Middlebrook 7H11 agar despite MG being a common adjunct of Middlebrook 7H11 agar modified for M. bovis growth. Subsequently, a selective MG-free Middlebrook 7H11 agar modified using haemolysed red cells and calf serum was demonstrated as the best performing (P < 0·05) medium for recovery of M. bovis from typical UK cheese types, Cheddar and Caerphilly. Significance and impact of the study: Following increased M. bovis infection of UK cattle, the risk posed to consumers from consumption of unpasteurized milk and dairy products has changed. Furthermore, published methods for the culture and molecular detection of M. bovis in raw milk products are limited. Cheese-making protocols and M. bovis culture media reported here provide tools for further investigation of M. bovis survival during all stages of cheese manufacture and could inform future assessment of the risk to consumers from M. bovis contamination of unpasteurized dairy products.
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Bacteriological agar, for molecular biology
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water, 0.2 μm filtered
Supelco
Nalidixic acid, analytical standard
Sigma-Aldrich
Nalidixic acid, ≥98%
Trimethoprim for system suitability, European Pharmacopoeia (EP) Reference Standard
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Trimethoprim, ≥98% (HPLC)
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Type A, ash 5-6%
Supelco
Trimethoprim, VETRANAL®, analytical standard
Sigma-Aldrich
Trimethoprim, crystallized, ≥99.0% (HPLC)
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type E, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Supelco
Trimethoprim, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Klebsiella Selective Supplement, suitable for microbiology
Sigma-Aldrich
Select agar
USP
Trimethoprim, United States Pharmacopeia (USP) Reference Standard
Trimethoprim, European Pharmacopoeia (EP) Reference Standard
Millipore
Agar, for chromogenic media