  • Search Results
  • Reduction of silver nanoparticles in DMF. Formation of monolayers and stable colloids.
Reduction of silver nanoparticles in DMF. Formation of monolayers and stable colloids.
Pure and Applied Chemistry. Chimie Pure Et Appliquee (2000)
Pastoriza S, et al.