  • Search Results
  • Particle size distribution and its relationship to black carbon in two urban and one rural site in Santiago de Chile.
Particle size distribution and its relationship to black carbon in two urban and one rural site in Santiago de Chile.
Journal of the Air & Waste Management Association (1995) (2014-08-16)
E Gramsch, E Reyes, P Oyola, Rubio Ma, G López, P Pérez, R Martínez
ABSTRACT
The size distribution of particles has been studied in three sites in the Metropolitan area of Santiago de Chile in the winter of 2009 and a comparison with black carbon was performed. Two sites are located near busy streets in Santiago and the other site is located in a rural area about 40 km west of Santiago with little influence from vehicles, but large influence from wood burning. The campaign lasted 1 or 2 weeks in each site. We have divided the particle size measurements into four groups (10-39 nm, 40-62 nm, 63-174 nm, and 175-700 nm) in order to compare with the carbon monitor. In the sites near the street, black carbon has a high correlation (R = 0.85) with larger particles (175-700 nm). The correlation decreased when black carbon was compared with smaller particles, having very small correlation with the smallest sizes (10-39 nm). In the rural site, black carbon also has a high correlation (R = 0.86) with larger particles (175-700 nm), but the correlation between black carbon and the finest particles (10-39 nm) decreases to near 0. These measurements are an indication that wood burning does not generate particles smaller than -50 nm. In the urban sites, particle size distribution is peaked toward smaller particles (10-39 nm) only during rush hours, but at other times, particles size distribution is peaked toward larger sizes. When solar radiation was high, evidence of secondary particle formation was seen in the rural site, but not in the urban sites. The correlation between the number of secondary particles and solar radiation was R2 = 0.46, indicating that it there may be other variables that play a role in ultrafine particle formation. Implications: A study of the size distribution of particles and black carbon concentration in two street sites and one rural site shows that in the last site the number of particles ultrafine particles (d < 40 nm) is 10 times lower but the number of larger particles is about 2 times lower. Thus, the rural site has less of the particles that are more dangerous to health. The number ofultrafine particles is mostly associated with traffic, while the number of larger particles is associated with wood burning and other sources. Wood burning does not generate particles smaller than -50 nm.
Product Description
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, puriss. p.a., powder
Carbon, foil, 10mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, tube, graphite, 50mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, foil, not light tested, 150x150mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, microleaf, 25x70mm, thinness 0.50μm, specific density 100μg/cm2, temporary glass support, annealed, 99.997%
Carbon, tube, graphite, 150mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 100mm, diameter 25mm, graphite, 99.95%
Carbon, tube, graphite, 100mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 100mm, diameter 5.0mm, graphite, 100%
Carbon, rod, 150mm, diameter 3.0mm, graphite, 100%
Carbon, rod, 150mm, diameter 50mm, graphite, 99.95%
Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%
Carbon, rod, 300mm, diameter 6.35mm, graphite, 100%
Carbon, tube, graphite, 100mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, tube, graphite, 50mm, outside diameter 12.7mm, inside diameter 9.5mm, wall thickness 1.6mm, 99.95%
Carbon, rod, 150mm, diameter 2.0mm, graphite, 100%
Carbon, foil, 5x10mm, thickness 2.0mm, hOpg
Carbon, foil, 250x250mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon, microleaf, 25x70mm, thinness 1.25μm, specific density 250μg/cm2, 99.997%