  • Search Results
  • The formation potential of haloacetonitriles in the Dez River water, Iran.
The formation potential of haloacetonitriles in the Dez River water, Iran.
Environmental technology (2014-08-26)
Mehdi Ahmadi, Bahman Ramavandi
ABSTRACT
The formation potential of haloacetonitriles (HANsFP) from chlorination of raw water of the Dez River in Iran was determined. Samples were collected before treatment at an intake of a water treatment plant. Tests were carried out to determine the effect of the reaction time (5-120 h), water pH (4-10), total organic carbon (TOC) content (2-8 mg L(-1)), chlorine dosage (3-25 mg L(-l)), water temperature (5-30 degrees C) and usual coagulants on HANsFP in the Dez River water. Increasing chlorine reaction time up to 80 h led to the remarkable increase in HANsFP and after that the HANsFP was gradually increased. Over 90% of the HANs was formed during 80 h reaction time. Linear relationships were found between the HANsFP and the chlorine dosage. The result showed that the HANsFP increased by increasing TOC content up to 6 mg L(-1), while further increases in the TOC level did not lead to any significant changes in the HANsFP level. The acidic water pH resulted in the highest HANsFP. An increase in water temperature enhanced the HANsFP; this factor should be considered for tropical waterworks. The ferric chloride had a high efficiency for the removal of HANsFP (72%). Overall, the Dez River water had a high potential for forming the HANs (approximately 17 microg L(-1)).
Product Description
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 30mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 50x50mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 6.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 10mm disks, thickness 0.2mm, glassy carbon
Carbon - Vitreous, foil, 8mm disks, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 7.0mm, glassy carbon
Carbon - Vitreous, foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon, foil, 100x100mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 100x100mm, thickness 0.125mm, rigid graphite, fine grain size, 99.95%
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon, foil, 100x100mm, thickness 0.25mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 100x100mm, thickness 0.35mm, flexible graphite, 99.8%
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 2.5mm, rigid graphite, fine grain size, 99.95%
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon
Carbon, foil, 100x100mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 10mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, tube, graphite, 50mm, outside diameter 3.18mm, inside diameter 1.18mm, wall thickness 1.0mm, 99.95%
Carbon, foil, not light tested, 150x150mm, thickness 0.075mm, flexible graphite, 99.8%