  • Search Results
  • Carbonic anhydrases, EPF2 and a novel protease mediate CO2 control of stomatal development.
Carbonic anhydrases, EPF2 and a novel protease mediate CO2 control of stomatal development.
Nature (2014-07-22)
Cawas B Engineer, Majid Ghassemian, Jeffrey C Anderson, Scott C Peck, Honghong Hu, Julian I Schroeder
ABSTRACT
Environmental stimuli, including elevated carbon dioxide levels, regulate stomatal development; however, the key mechanisms mediating the perception and relay of the CO2 signal to the stomatal development machinery remain elusive. To adapt CO2 intake to water loss, plants regulate the development of stomatal gas exchange pores in the aerial epidermis. A diverse range of plant species show a decrease in stomatal density in response to the continuing rise in atmospheric CO2 (ref. 4). To date, one mutant that exhibits deregulation of this CO2-controlled stomatal development response, hic (which is defective in cell-wall wax biosynthesis, ref. 5), has been identified. Here we show that recently isolated Arabidopsis thaliana β-carbonic anhydrase double mutants (ca1 ca4) exhibit an inversion in their response to elevated CO2, showing increased stomatal development at elevated CO2 levels. We characterized the mechanisms mediating this response and identified an extracellular signalling pathway involved in the regulation of CO2-controlled stomatal development by carbonic anhydrases. RNA-seq analyses of transcripts show that the extracellular pro-peptide-encoding gene EPIDERMAL PATTERNING FACTOR 2 (EPF2), but not EPF1 (ref. 9), is induced in wild-type leaves but not in ca1 ca4 mutant leaves at elevated CO2 levels. Moreover, EPF2 is essential for CO2 control of stomatal development. Using cell-wall proteomic analyses and CO2-dependent transcriptomic analyses, we identified a novel CO2-induced extracellular protease, CRSP (CO2 RESPONSE SECRETED PROTEASE), as a mediator of CO2-controlled stomatal development. Our results identify mechanisms and genes that function in the repression of stomatal development in leaves during atmospheric CO2 elevation, including the carbonic-anhydrase-encoding genes CA1 and CA4 and the secreted protease CRSP, which cleaves the pro-peptide EPF2, in turn repressing stomatal development. Elucidation of these mechanisms advances the understanding of how plants perceive and relay the elevated CO2 signal and provides a framework to guide future research into how environmental challenges can modulate gas exchange in plants.
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Endoproteinase Arg-C from mouse submaxillary gland, suitable for protein sequencing, lyophilized powder
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder