  • Search Results
  • Epigenomic characterization of locally advanced anal cancer: a radiation therapy oncology group 98-11 specimen study.
Epigenomic characterization of locally advanced anal cancer: a radiation therapy oncology group 98-11 specimen study.
Diseases of the colon and rectum (2014-07-09)
Erin M Siegel, Steven Eschrich, Kathryn Winter, Bridget Riggs, Anders Berglund, Abidemi Ajidahun, Jeff Simko, Jennifer Moughan, Jaffer Ajani, Anthony Magliocco, Abul Elahi, Sarah Hoffe, David Shibata
ABSTRACT
The Radiation Therapy Oncology Group 98-11 clinical trial demonstrated the superiority of standard 5-fluorouracil/mitomycin-C over 5-fluorouracil/cisplatin in combination with radiation in the treatment of anal squamous cell cancer. Tumor size (>5 cm) and lymph node metastases are associated with disease progression. There may be key molecular differences (eg, DNA methylation changes) in tumors at high risk for progression. The objectives of this study were to determine whether there are differences in DNA methylation at individual CpG sites and within genes among locally advanced anal cancers, with large tumor size and/or nodal involvement, compared with those that are less advanced. This was a case-case study among 121 patients defined as high risk (tumor size >5 cm and/or nodal involvement; n = 59) or low risk (≤5 cm, node negative; n = 62) within the mitomycin-C arm of the Radiation Therapy Oncology Group 98-11 trial. DNA methylation was measured using the Illumina HumanMethylation450 Array. The study was conducted in a tertiary care cancer center in collaboration with a national clinical trials cooperative group. The patients consisted of 74 women and 47 men with a median age of 54 years (range, 25-79 years). DNA methylation differences at individual CpG sites and within genes between low- and high-risk patients were compared using the Mann-Whitney test (p < 0.001). A total of 16 CpG loci were differentially methylated (14 increased and 2 decreased) in high- versus low-risk cases. Genes harboring differentially methylated CpG sites included known tumor suppressor genes and novel targets. This study only included patients in the mitomycin-C arm with tumor tissue; however, this sample was representative of the trial. This is the first study to apply genome-wide methylation analysis to anal cancer. Biologically relevant differences in methylated targets were found to discriminate locally advanced from early anal cancer. Epigenetic events likely play a significant role in the progression of anal cancer and may serve as potential biomarkers.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥98% (HPLC), potency: ≥970 μg per mg (USP XXIV), γ-irradiated, suitable for cell culture
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, ≥970 μg/mg (USP XXIV)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Mitomycin, European Pharmacopoeia (EP) Reference Standard
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
5-Fluorouracil, analytical standard