  • Search Results
  • Why selenocysteine replaces cysteine in thioredoxin reductase: a radical hypothesis.
Why selenocysteine replaces cysteine in thioredoxin reductase: a radical hypothesis.
Biochemistry (2014-07-08)
Thomas Nauser, Daniel Steinmann, Guido Grassi, Willem H Koppenol
ABSTRACT
Thioredoxin reductases, important biological redox mediators for two-electron transfers, contain either 2 cysteines or a cysteine (Cys) and a selenocysteine (Sec) at the active site. The incorporation of Sec is metabolically costly, and therefore surprising. We provide here a rationale: in the case of an accidental one-electron transfer to a S-S or a S-Se bond during catalysis, a thiyl or a selanyl radical, respectively would be formed. The thiyl radical can abstract a hydrogen from the protein backbone, which subsequently leads to the inactivation of the protein. In contrast, a selanyl radical will not abstract a hydrogen. Therefore, formation of Sec radicals in a GlyCysSecGly active site will less likely result in the destruction of a protein compared to a GlyCysCysGly active site.
Product Description
Selenium, pellets, < 5mm, ≥99.999%
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Cysteine, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Supelco
Sulfur, PESTANAL®, analytical standard
SAFC
L-Cysteine
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Sulfur, 99.998% trace metals basis
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
L-Cysteine, BioUltra, ≥98.5% (RT)
Supelco
L-Cysteine, certified reference material, TraceCERT®
Sigma-Aldrich
L-Cysteine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0%