  • Search Results
  • Detection and molecular cloning of CYP74Q1 gene: identification of Ranunculus acris leaf divinyl ether synthase.
Detection and molecular cloning of CYP74Q1 gene: identification of Ranunculus acris leaf divinyl ether synthase.
Biochimica et biophysica acta (2014-05-28)
Svetlana S Gorina, Yana Y Toporkova, Lucia S Mukhtarova, Ivan R Chechetkin, Bulat I Khairutdinov, Yuri V Gogolev, Alexander N Grechkin
ABSTRACT
Enzymes of the CYP74 family, including the divinyl ether synthase (DES), play important roles in plant cell signalling and defence. The potent DES activities have been detected before in the leaves of the meadow buttercup (Ranunculus acris L.) and few other Ranunculaceae species. The nature of these DESs and their genes remained unrevealed. The PCR with degenerate primers enabled to detect the transcript of unknown P450 gene assigned as CYP74Q1. Besides, two more CYP74Q1 isoforms with minimal sequence variations have been found. The full length recombinant CYP74Q1 protein was expressed in Escherichia coli. The preferred substrates of this enzyme are the 13-hydroperoxides of α-linolenic and linoleic acids, which are converted to the divinyl ether oxylipins (ω5Z)-etherolenic acid, (9Z,11E)-12-[(1'Z,3'Z)-hexadienyloxy]-9,11-dodecadienoic acid, and (ω5Z)-etheroleic acid, (9Z,11E)-12-[(1'Z)-hexenyloxy]-9,11-dodecadienoic acid, respectively, as revealed by the data of mass spectrometry, NMR and UV spectroscopy. Thus, CYP74Q1 protein was identified as the R. acris DES (RaDES), a novel DES type and the opening member of new CYP74Q subfamily.
Product Description
USP
Hypromellose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Supelco
Linolenic acid, analytical standard