  • Search Results
  • Assessment of the potential of polyphenols as a CYP17 inhibitor free of adverse corticosteroid elevation.
Assessment of the potential of polyphenols as a CYP17 inhibitor free of adverse corticosteroid elevation.
Biochemical pharmacology (2014-05-31)
Chun-Ju Lin, Li-Chuan Cheng, Tsu-Chun Emma Lin, Chien-Jen Wang, Lih-Ann Li
ABSTRACT
Inhibition of 17α-hydroxylase/17,20-lyase (CYP17), which dictates the proceeding of androgen biosynthesis, is recommended as an effective treatment for androgen-dependent diseases. However, androgen depletion by selective CYP17 inhibition is accompanied with corticosteroid elevation, which increases risk of cardiovascular diseases. In this study, we evaluated the likelihood of polyphenols as a CYP17 inhibitor without cardiovascular complications. All examined polyphenols significantly inhibited CYP17 in human adrenocortical H295R cells, but their effects on androgen and cortisol biosynthesis were diverse. Resveratrol was the most potent CYP17 inhibitor with an approximate IC50 of 4 μM, and the inhibition might weigh on the 17α-hydroxylase activity more than the 17,20-lyase activity. Resveratrol also inhibited 21α-hydroxylase (CYP21) essential for corticosteroid biosynthesis but to a lesser extent, thus preventing the occurrence of cortisol elevation following CYP17 blockade. Although transcriptional down-regulation was important for α-naphthoflavone-mediated CYP17 inhibition, resveratrol inhibited CYP17 and CYP21 mainly at the level of enzyme activity rather than enzyme abundance and cytochrome P450 electron transfer. Daidzein also inhibited CYP17 and CYP21 although less potent than resveratrol. Daidzein was the only polyphenol showing inhibition of 3β-hydroxysteroid dehydrogenase type II (3βHSD2). The exceptional 3βHSD2 inhibition led to dehydroepiandrosterone accumulation alongside daidzein-caused androgen biosynthetic impairment. In contrast, androgen and cortisol secretion was increased or remained normal under α-naphthoflavone and β-naphthoflavone treatments, suggesting that CYP17 inhibition was counteracted by increased substrate generation. α-naphthoflavone and β-naphthoflavone also enhanced the formation of cortisol from 17-hydroxyprogesterone and testosterone from androstenedione. Our findings suggest a potential application of resveratrol in androgen deprivation therapy.
Product Description
Supelco
Resveratrol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Phenol Red, ACS reagent
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Supelco
Dehydroepiandrosterone (DHEA) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)