  • Search Results
  • Unusual penetration of phospholipid mono- and bilayers by Quillaja bark saponin biosurfactant.
Unusual penetration of phospholipid mono- and bilayers by Quillaja bark saponin biosurfactant.
Biochimica et biophysica acta (2014-04-22)
Kamil Wojciechowski, Marta Orczyk, Thomas Gutberlet, Marcus Trapp, Kuba Marcinkowski, Tomasz Kobiela, Thomas Geue
ABSTRACT
The interactions between a model phospholipid 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) and a biosurfactant Quillaja Bark Saponin (QBS) obtained from the bark of Quillaja saponaria Molina were studied using simple models of biological membranes. QBS is known to interact strongly with the latter, exerting a number of haemolytic, cytotoxic and anti-microbial actions. The interaction of QBS dissolved in the subphase with DPPC monolayers and silicon-supported bilayers was studied above the cmc (10(-3)M). Surface pressure relaxation and surface dilatational rheology combined with quartz crystal microbalance (QCM) and neutron reflectivity (NR) were employed for this purpose. The DPPC-penetrating abilities of QBS are compared with those of typical synthetic surfactants (SDS, CTAB and Triton X-100). We show that the penetration studies using high surface activity (bio)surfactants should be performed by a subphase exchange, not by spreading onto the surfactant solution. In contrast to the synthetic surfactants of similar surface activity, QBS does not collapse DPPC mono- and bilayers, but penetrates them, improving their surface dilatational elastic properties even in the highly compressed solid state. The dilatational viscoelasticity modulus increases from 204 mN/m for pure DPPC up to 310 mN/m for the QBS-penetrated layers, while it drops to near zero values in the case of the synthetic surfactants. The estimated maximum insertion pressure of QBS into DPPC monolayers exceeds the maximum surface pressure achievable in our setup, in agreement with the surface rheological response of the penetrated layers.
Product Description
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
SAFC
Hexadecyltrimethylammonium bromide
Supelco
Acetone solution, contains 20.0 % (v/v) acetonitrile, suitable for HPLC
Supelco
Acetone solution, certified reference material, 2000 μg/mL in methanol: water (9:1)
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, semisynthetic, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Supelco
Acetone, analytical standard
Sigma-Aldrich
1,2-Dipalmitoyl-rac-glycero-3-phosphocholine, ~99%
Sigma-Aldrich
Saponin, used as non-ionic surfactant
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard