  • Search Results
  • Hydroxytyrosol ameliorates oxidative stress and mitochondrial dysfunction in doxorubicin-induced cardiotoxicity in rats with breast cancer.
Hydroxytyrosol ameliorates oxidative stress and mitochondrial dysfunction in doxorubicin-induced cardiotoxicity in rats with breast cancer.
Biochemical pharmacology (2014-04-15)
Sergio Granados-Principal, Nuri El-Azem, Reinald Pamplona, Cesar Ramirez-Tortosa, Mario Pulido-Moran, Laura Vera-Ramirez, Jose L Quiles, Pedro Sanchez-Rovira, Alba Naudí, Manuel Portero-Otin, Patricia Perez-Lopez, Mcarmen Ramirez-Tortosa
ABSTRACT
Oxidative stress is involved in several processes including cancer, aging and cardiovascular disease, and has been shown to potentiate the therapeutic effect of drugs such as doxorubicin. Doxorubicin causes significant cardiotoxicity characterized by marked increases in oxidative stress and mitochondrial dysfunction. Herein, we investigate whether doxorubicin-associated chronic cardiac toxicity can be ameliorated with the antioxidant hydroxytyrosol in rats with breast cancer. Thirty-six rats bearing breast tumors induced chemically were divided into 4 groups: control, hydroxytyrosol (0.5mg/kg, 5days/week), doxorubicin (1mg/kg/week), and doxorubicin plus hydroxytyrosol. Cardiac disturbances at the cellular and mitochondrial level, mitochondrial electron transport chain complexes I-IV and apoptosis-inducing factor, and oxidative stress markers have been analyzed. Hydroxytyrosol improved the cardiac disturbances enhanced by doxorubicin by significantly reducing the percentage of altered mitochondria and oxidative damage. These results suggest that hydroxytyrosol improve the mitochondrial electron transport chain. This study demonstrates that hydroxytyrosol protect rat heart damage provoked by doxorubicin decreasing oxidative damage and mitochondrial alterations.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Supelco
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
Sigma-Aldrich
3-Hydroxytyrosol, ≥98% (HPLC)
Supelco
Phenylethyl Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol
Sigma-Aldrich
Osmium tetroxide solution, 4 wt. % in H2O
Sigma-Aldrich
Phenethyl alcohol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Phenethyl alcohol, ≥99%, FCC, FG
Sigma-Aldrich
2-Phenylethanol, ≥99.0% (GC)