  • Search Results
  • Dissipation kinetics and effect of different decontamination techniques on the residues of emamectin benzoate and spinosad in cowpea pods.
Dissipation kinetics and effect of different decontamination techniques on the residues of emamectin benzoate and spinosad in cowpea pods.
Environmental monitoring and assessment (2014-04-01)
V Vijayasree, Hebsy Bai, Thomas Biju Mathew, Thomas George, George Xavier, N Pratheesh Kumar, S Visalkumar
ABSTRACT
Dissipation and decontamination of the semisynthetic macrolide emamectin benzoate and the natural insecticide spinosad on cowpea pods were studied following field application at single and double doses of 11.0 and 22 and 73 and 146 g ai ha(-1), respectively. Residues of these naturalytes were estimated using LC-MS/MS. The initial deposit of 0.073 and 0.153 mg kg(-1) of emamectin benzoate dissipated below quantitation level on the fifth and seventh day at single and double dosage, respectively. For spinosad, the initial deposits of 0.94 and 1.90 mg kg(-1) reached below quantitation level on the 7th day and 15th day at single and double dosage, respectively. The half-life of emamectin benzoate and spinosad was 1.13-1.49 and 1.05-1.39 days with the calculated safe waiting period of 2.99-6.12 and 1.09-3.25 days, respectively, for single and double dosage. Processing of the harvestable pods with different decontamination techniques resulted in 33.82 to 100 % removal 2 h after the application of emamectin benzoate and 100 % removal 3 days after spraying, while the removal was 42.05 to 87.46 % 2 h after the application of spinosad and 38.05 to 68.08 % 3 days after application.
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Ivermectin
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Supelco
Acetonitrile, HPLC grade, ≥99.93%
Supelco
Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Ivermectin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)