  • Search Results
  • Fast disintegrating tablets of nisoldipine for intra-oral administration.
Fast disintegrating tablets of nisoldipine for intra-oral administration.
Pharmaceutical development and technology (2013-07-12)
Gamal M El Maghraby, Ramy N Elsergany
ABSTRACT
Nisoldipine is a calcium channel blocker with low and variable oral bioavailability. This was attributed to slow dissolution and presystemic metabolism. Accordingly, the objective of this work was to enhance the dissolution rate of nisoldipine to formulate fast disintegrating tablets with rapid dissolution. Binary solid dispersions (SD) were prepared for the drug with hydroxypropyl methyl cellulose E5 (HPMC), polyvinylpyrrolidone (PVP), Pluronic F68 or polyethylene glycol 6000 (PEG 6000). SD formation increased the dissolution rate compared to pure drug with the corresponding physical mixtures failing to provide the same dissolution enhancement. This indicates that the SD enhanced dissolution is not due to the solubilizing effect of the polymer and can be due to physical change in the drug crystal which was confirmed by thermal analysis. SD with HPMC and PVP were selected for preparation of fast disintegrating tablets as they liberated most of the drug in the first 5 min. HPMC-based tablets disintegrated rapidly and released most of the drug in the first 2 min which correlated with the corresponding SD. In contrast, PVP-based tablets disintegrated slowly with gradual dissolution. This can be attributed to the binding effect of PVP. The study developed fast disintegrating tablet for intra-oral administration.
Sigma-Aldrich
Methyl cellulose, viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Povidone, meets USP testing specifications
Sigma-Aldrich
Methocel® A15C, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Kollidon® 25
Supelco
Poly(vinylpolypyrrolidone), ~110 μm particle size
Sigma-Aldrich
Methocel® A4M, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Supelco
Povidone, Pharmaceutical Secondary Standard; Certified Reference Material
Crospovidone, European Pharmacopoeia (EP) Reference Standard
USP
Crospovidone, United States Pharmacopeia (USP) Reference Standard
USP
Povidone, United States Pharmacopeia (USP) Reference Standard
Povidone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Polyvinylpyrrolidone, average mol wt 40,000
Sigma-Aldrich
Polyvinylpyrrolidone, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Polyvinylpyrrolidone, for molecular biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
Polyvinylpyrrolidone, K 30
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~29,000
Sigma-Aldrich
Polyvinylpyrrolidone, powder, average Mw ~55,000
Sigma-Aldrich
Polyvinylpyrrolidone, average Mw ~1,300,000 by LS
Sigma-Aldrich
Polyvinylpyrrolidone, K 90
Sigma-Aldrich
Methocel® A15 LV, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Polyvinylpyrrolidone solution, K 60, 45% in H2O