MilliporeSigma
  • Multimodality therapy and liver transplantation for hepatocellular carcinoma: a 14-year prospective analysis of outcomes.
Multimodality therapy and liver transplantation for hepatocellular carcinoma: a 14-year prospective analysis of outcomes.
Transplantation (2014-02-08)
Rajesh Ramanathan, Amit Sharma, David D Lee, Martha Behnke, Karen Bornstein, R Todd Stravitz, Malcolm Sydnor, Ann Fulcher, Adrian Cotterell, Marc P Posner, Robert A Fisher
ABSTRACT
Hepatocellular carcinoma is a major cause of death among patients with cirrhosis. A standardized approach of multimodality therapy with intent-to-treat by transplantation for all patients with hepatocellular carcinoma was instituted at our transplant center in 1997. Data were prospectively collected to evaluate the impact of multimodality therapy on posttransplant patient survival, tumor recurrence, and patient survival without transplantation. All patients with hepatocellular carcinoma were eligible for multimodality therapy. Multimodality therapy consisted of hepatic resection, radiofrequency ablation, transarterial chemoembolization, transarterial chemoinfusion, yttrium-90 microsphere radioembolization, and sorafenib. Approximately 715 patients underwent multimodality therapy; 231 patients were included in the intent-to-treat with transplantation arm, and 484 patients were treated with multimodality therapy or palliative therapy because of contraindications for transplantation. A 60.2% transplantation rate was achieved in the intent-to-treat with transplantation arm. Posttransplant survivals at 1 and 5 years were 97.1% and 72.5%, respectively. Tumor recurrence rates at 1, 3, and 5 years were 2.4%, 6.2%, and 11.6%, respectively. Patients with contraindications to transplant had increased 1- and 5-year survival from diagnosis with multimodality therapy compared with those not treated (73.1% and 46.5% versus 15.5% and 4.4%, P<0.0001). Using multimodality therapy before liver transplantation for hepatocellular carcinoma achieved low recurrence rates and posttransplant survival equivalent to patients with primary liver disease without hepatocellular carcinoma. Multimodality therapy may help identify patients with less active tumor biology and result in improved disease-free survival and organ utilization.
Product Description
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Supelco
Nicotinamide (Niacinamide), analytical standard