  • Search Results
  • [Theoretical, methodological and clinical aspects of the growth of granulopoietic cells in agar culture].
[Theoretical, methodological and clinical aspects of the growth of granulopoietic cells in agar culture].
Folia haematologica (Leipzig, Germany : 1928) (1978-01-01)
W Schultze, W Helbig
PMID77816
ABSTRACT
Haematopoietic stem cells are capable of developing colonies of differentiated granulocytes in a semi-stable agar environment under corresponding experimental conditions. Number and size of aggregations developing from a single stem cell in each case called "in culture Colony Forming Unit (CFU-c)" enable conclusions to be made about the functional ability of the cultivated human or animal bone marrow. The method is applied in investigating the kinetics in the haematopoietic system and, in addition, it is used for diagnostics, therapy and control of the course in haematological diseases as well as to check the proliferous ability of cryopreserved bone marrow cells. As to the standardization of the procedure the instability of the active principle of the foetal calf serum as well as the essential colony stimulating factor represent limiting items at present.
Sigma-Aldrich
Bacteriological agar, for molecular biology
Sigma-Aldrich
Select agar
Millipore
Agar, for chromogenic media
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, Type A, ash 5-6%
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type E, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Millipore
Agar, high purity, low ionic content, low gel strenght
Millipore
Agar, for membrane filtration, low gel strengh