  • Search Results
  • Microbial modulators of soil carbon storage: integrating genomic and metabolic knowledge for global prediction.
Microbial modulators of soil carbon storage: integrating genomic and metabolic knowledge for global prediction.
Trends in microbiology (2013-10-22)
Pankaj Trivedi, Ian C Anderson, Brajesh K Singh
ABSTRACT
Soil organic carbon performs a number of functions in ecosystems and it is clear that microbial communities play important roles in land-atmosphere carbon (C) exchange and soil C storage. In this review, we discuss microbial modulators of soil C storage, 'omics'-based approaches to characterize microbial system interactions impacting terrestrial C sequestration, and how data related to microbial composition and activities can be incorporated into mechanistic and predictive models. We argue that although making direct linkage of genomes to global phenomena is a significant challenge, many connections at intermediate scales are viable with integrated application of new systems biology approaches and powerful analytical and modelling techniques. This integration could enhance our capability to develop and evaluate microbial strategies for capturing and sequestering atmospheric CO2.
Product Description
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Sigma-Aldrich
Activated charcoal, DARCO® KB-G
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, acid-washed with phosphoric and sulfuric acids
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, 4-14 mesh, granular, Norit® PK 3-5
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder
Sigma-Aldrich
Activated charcoal, Norit® ROW 0.8 SUPRA, pellets
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Carbon, tube, graphite, 150mm, outside diameter 6.35mm, inside diameter 3.15mm, wall thickness 1.6mm, 99.95%
Carbon - Vitreous, rod, 100mm, diameter 7.0mm, glassy carbon