Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Supelco
Activated charcoal, suitable for GC
Supelco
Activated charcoal, powder
Carbon, foil, not light tested, 300x300mm, thickness 0.075mm, flexible graphite, 99.8%
Carbon, foil, 250x250mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 150x150mm, thickness 0.5mm, flexible graphite, 99.8%
Carbon, foil, 25x25mm, thickness 0.1mm, pyrolytic graphite, 99.99%
Carbon, foil, 500x500mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 500x500mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, microleaf, 25x70mm, thinness 2.5μm, specific density 500μg/cm2, 99.997%
Carbon, foil, 50x50mm, thickness 0.5mm, rigid graphite, fine grain size, 99.997%
Carbon, foil, 50x50mm, thickness 0.1mm, pyrolytic graphite, 99.99%
Carbon, rod, 50mm, diameter 25mm, graphite, 100%
Carbon, foil, 25x25mm, thickness 0.5mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 300x300mm, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 100mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 15mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 50x50mm, thickness 1.0mm, rigid graphite, fine grain size, 99.95%
Carbon, foil, 50x50mm, thickness 5.0mm, rigid graphite, medium grain size, 99.5%
Carbon, foil, 5x10mm, thickness 2.0mm, hOpg
Carbon, microleaf, 25x70mm, thinness 0.50μm, specific density 100μg/cm2, temporary glass support, 99.997%
Carbon, rod, 300mm, diameter 3.0mm, graphite, 100%
Carbon, rod, 300mm, diameter 5.0mm, graphite, 100%
Carbon, foil, 50x50mm, thickness 1.0mm, flexible graphite, 99.8%
Carbon, foil, 4mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 6mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, foil, 8mm disks, thickness 0.2mm, flexible graphite, 99.8%
Carbon, rod, 100mm, diameter 25mm, graphite, 100%