MilliporeSigma
Lamotrigine (Lamictal IR) for the treatment of bipolar disorder.
Expert opinion on pharmacotherapy (2012-11-13)
Charles L Bowden, Vivek Singh
ABSTRACT
Over the past decade the use of lamotrigine in bipolar disorder has increased. However, the evidence base suggests a more limited role for lamotrigine as part of an overall treatment regimen in bipolar disorder. We reviewed publications of randomized clinical trials of lamotrigine, emphasizing studies in bipolar disorder. The low burden of adverse effects with lamotrigine has been confirmed in these studies. Its lack of benefit in acute mania is established. Despite modest benefits for a subset of depressive episodes in bipolar disorder, it was not superior to placebo in well-designed studies. As monotherapy, in randomized, blinded trials in rapid cycling bipolar disorder it was not superior to placebo. Its role in maintenance treatment is relatively well established as one component of combination therapy, with evidence for benefits when combined with lithium or valproate. Combination regimens including lamotrigine appear to be superior to monotherapy. Monotherapy utilization of lamotrigine for maintenance treatment is not supported by these studies. Lamotrigine has benefits in bipolar disorder management principally as a component of combination treatment which includes a mood stabilizer. The utility of lamotrigine in acute bipolar depression and major depressive disorder is modest.
Sigma-Aldrich
Lamotrigine, ≥98%, powder
Lamotrigine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Lamotrigine, United States Pharmacopeia (USP) Reference Standard
Lamotrigine for peak identification, European Pharmacopoeia (EP) Reference Standard
Lamotrigine, European Pharmacopoeia (EP) Reference Standard
Supelco
Lamotrigine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Lamotrigine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®