  • Search Results
  • Skin antisepsis: using 2% chlorhexidine gluconate in 70% isopropyl alcohol.
Skin antisepsis: using 2% chlorhexidine gluconate in 70% isopropyl alcohol.
British journal of nursing (Mark Allen Publishing) (2008-03-26)
Sheila Inwood
ABSTRACT
The insertion of peripheral and central venous access devices carries a high risk of healthcare-associated infection. One of the main risks of this procedure is the introduction of microorganisms from the patient's own skin. To combat this, and to reduce the rates of healthcare-associated infection, effective skin antisepsis using 2% chlorhexidine gluconate (CHG) in 70% isopropyl alcohol (IPA) has been recommended by the epic2: National Evidence-Based Guidelines for Preventing Healthcare-Associated Infections in NHS Hospitals in England (Pratt et al, 2007). This article discribes the introduction of 2% CHG in 70% IPA into the United Kingdom healthcare market and gives a discussion of the recommendations of epic2 (Pratt et al, 2007). The article identifies the correct use of 2%CHG 70% IPA solution, by detailing the variety of products and applications now available. It also clarifies their correct application and use to enable Trusts to provide a high standard of infection control and reduce healthcare-associated infection. It highlights the potential incorrect use of the available products due to cost pressures within the healthcare setting.
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Isopropanol, 70% in H2O
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, 99.5%, HPLC grade
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard