  • Search Results
  • ACOG Committee Opinion no. 594: Immersion in water during labor and delivery.
ACOG Committee Opinion no. 594: Immersion in water during labor and delivery.
Obstetrics and gynecology (2014-05-03)
ABSTRACT
Immersion in water has been suggested as a beneficial alternative for labor, or delivery, or both and over the past decades has gained popularity in many parts of world. Immersion in water during the first stage of labor may be associated with decreased pain or use of anesthesia and decreased duration of labor. However, there is no evidence that immersion in water during the first stage of labor otherwise improves perinatal outcomes, and it should not prevent or inhibit other elements of care. The safety and efficacy of immersion in water during the second stage of labor have not been established, and immersion in water during the second stage of labor has not been associated with maternal or fetal benefit. Given these facts and case reports of rare but serious adverse effects in the newborn, the practice of immersion in the second stage of labor (underwater delivery) should be considered an experimental procedure that only should be performed within the context of an appropriately designed clinical trial with informed consent. Facilities that plan to offer immersion in the first stage of labor need to establish rigorous protocols for candidate selection, maintenance and cleaning of tubs and immersion pools, infection control procedures, monitoring of mothers and fetuses at appropriate intervals while immersed, and immediately and safely moving women out of the tubs if maternal or fetal concerns develop.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Supelco
Water, for TOC analysis
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, PCR Reagent
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, BioPerformance Certified
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, suitable for HPLC