  • Search Results
  • Heat removal using microclimate foot cooling: a thermal foot manikin study.
Heat removal using microclimate foot cooling: a thermal foot manikin study.
Aviation, space, and environmental medicine (2014-04-24)
John W Castellani, Robert Demes, Thomas L Endrusick, Samuel N Cheuvront, Scott J Montain
ABSTRACT
It has been proposed that microclimate cooling systems exploit the peripheral extremities because of more efficient heat transfer. The purpose of this study was to quantify, using a patented microclimate cooling technique, the heat transfer from the plantar surface of the foot for comparison to other commonly cooled body regions. A military boot was fitted with an insole embedded with a coiled, 1.27 m length of hollow tubing terminating in inlet and outlet valves. A thermal foot manikin with a surface temperature of 34 degrees C was placed in the boot and the valves were connected to a system that circulated water through the insole at a temperature of 20 degrees C and flow rate of 120 ml x min(-1). The manikin foot served as a constant heat source to determine heat transfer provided by the insole. Testing was done with the foot model dry and sweating at a rate of 500 ml x h(- 1) x m(-2). Climatic chamber conditions were 30 degrees C with 30% RH. Heat loss was approximately 4.1 +/- 0.1 and approximately 7.7 +/- 0.3 W from the dry and sweating foot models, respectively. On a relative scale, the heat loss was 3.0 W and 5.5 W per 1% (unit) body surface area, respectively, for the dry and sweating conditions. The relative heat loss afforded by plantar foot cooling was similar compared to other body regions, but the absolute amount of heat removal is unlikely to make an impact on whole body heat balance.
Sigma-Aldrich
Water, deionized
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
Water, HPLC Plus
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O