  • Search Results
  • Preparation and characterization of fullerene (C60) amino acid nanoparticles for liver cancer cell treatment.
Preparation and characterization of fullerene (C60) amino acid nanoparticles for liver cancer cell treatment.
Journal of nanoscience and nanotechnology (2014-04-18)
Zhi Li, Li-Li Pan, Fei-Long Zhang, Zhiyuan Wang, Ying-Ying Shen, Zhen-Zhong Zhang
ABSTRACT
The properties of an ideal photosensitizer are water solubility, low cytotoxicity in the dark, high ability to produce reactive oxygen species (ROS). The characteristics of water-soluble fullerene (C60) amino acid nanoparticles as a photosensitizer were evaluated. C60 modified with l-phenylalanine (C60-phe) or glycine (C60-gly) was very efficient to carry out photodynamic activity leading to cleavage of plasmid DNA in vitro. These C60 amino acid nanoparticles were the most active photosensitizer against human Liver cancer cells and induced cancer cells apoptosis after illumination. However, these derivatives exhibited no significant cytotoxicity in dark. It produced diffuse intracellular fluorescence when 2',7'-dichlorfluorescein-diacetate (DCFH-DA) was added as an ROS probe, suggesting phototoxicity of these derivatives related with the generation of intracellular ROS. These findings indicate that these fullerene derivatives may be excellent candidate PDT enhancing agents.
Product Description
Supelco
L-Phenylalanine, certified reference material, TraceCERT®
Sigma-Aldrich
Fullerene-C60, sublimed, 99.9%
Supelco
L-Phenylalanine, analytical standard, for Nitrogen Determination According to Kjeldahl Method
Sigma-Aldrich
Fullerene-C60, 99.5%
Sigma-Aldrich
L-Phenylalanine, 99%, FCC
Sigma-Aldrich
Fullerene-C60, 98%
Supelco
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
USP
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
Phenylalanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine hydrochloride, ≥99% (HPLC)
Sigma-Aldrich
L-Phenylalanine, reagent grade, ≥98%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
SAFC
Glycine
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Glycine, European Pharmacopoeia (EP) Reference Standard
USP
Glycine, United States Pharmacopeia (USP) Reference Standard