  • Search Results
  • Ocular-specific ER stress reduction rescues glaucoma in murine glucocorticoid-induced glaucoma.
Ocular-specific ER stress reduction rescues glaucoma in murine glucocorticoid-induced glaucoma.
The Journal of clinical investigation (2014-04-03)
Gulab S Zode, Arti B Sharma, Xiaolei Lin, Charles C Searby, Kevin Bugge, Gun Hee Kim, Abbot F Clark, Val C Sheffield
ABSTRACT
Administration of glucocorticoids induces ocular hypertension in some patients. If untreated, these patients can develop a secondary glaucoma that resembles primary open-angle glaucoma (POAG). The underlying pathology of glucocorticoid-induced glaucoma is not fully understood, due in part to lack of an appropriate animal model. Here, we developed a murine model of glucocorticoid-induced glaucoma that exhibits glaucoma features that are observed in patients. Treatment of WT mice with topical ocular 0.1% dexamethasone led to elevation of intraocular pressure (IOP), functional and structural loss of retinal ganglion cells, and axonal degeneration, resembling glucocorticoid-induced glaucoma in human patients. Furthermore, dexamethasone-induced ocular hypertension was associated with chronic ER stress of the trabecular meshwork (TM). Similar to patients, withdrawal of dexamethasone treatment reduced elevated IOP and ER stress in this animal model. Dexamethasone induced the transcriptional factor CHOP, a marker for chronic ER stress, in the anterior segment tissues, and Chop deletion reduced ER stress in these tissues and prevented dexamethasone-induced ocular hypertension. Furthermore, reduction of ER stress in the TM with sodium 4-phenylbutyrate prevented dexamethasone-induced ocular hypertension in WT mice. Our data indicate that ER stress contributes to glucocorticoid-induced ocular hypertension and suggest that reducing ER stress has potential as a therapeutic strategy for treating glucocorticoid-induced glaucoma.
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sodium phenylbutyrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
4-Phenylbutyric acid, 99%
Sigma-Aldrich
Sodium phenylbutyrate, ≥98% (HPLC)
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard