  • Search Results
  • Role of cell cycle regulatory molecules in retinoic acid- and vitamin D3-induced differentiation of acute myeloid leukaemia cells.
Role of cell cycle regulatory molecules in retinoic acid- and vitamin D3-induced differentiation of acute myeloid leukaemia cells.
Cell proliferation (2014-03-22)
X T Hu, K S Zuckerman
ABSTRACT
The important role of cell cycle regulatory molecules in all trans-retinoic acid (ATRA)- and vitamin D3-induced growth inhibition and differentiation induction has been intensively studied in both acute myeloid leukaemia primary cells and a variety of leukaemia cell lines. Cyclin-dependent kinases (CDK)-activating kinase has been demonstrated to interact with retinoic acid receptor (RAR)α in acute promyelocytic leukaemia cells, and inhibition of CDK-activating kinase by ATRA causes hypophosphorylation of PML-RARα, leading to myeloid differentiation. In many cases, downregulation of CDK activity by ATRA and vitamin D3 is a result of elevated p21- and p27-bound CDKs. Activation of p21 is regulated at the transcriptional level, whereas elevated p27 results from both (indirectly) transcriptional activation and post-translational modifications. CDK inhibitors (CKIs) of the INK family, such as p15, p16 and p18, are mainly involved in inhibition of cell proliferation, whereas CIP/KIP members, such as p21, regulate both growth arrest and induction of differentiation. ATRA and vitamin D3 can also downregulate expression of G1 CDKs, especially CDK2 and CDK6. Inhibition of cyclin E expression has only been observed in ATRA- but not in vitamin D3-treated leukaemic cells. In vitro, not only dephosphorylation of pRb but also elevation of total pRb is required for ATRA and vitamin D3 to suppress growth and trigger their differentiation. Finally, sharp reduction in c-Myc has been observed in several leukaemia cell lines treated with ATRA, which may regulate expression of CDKs and CKIs.
Sigma-Aldrich
Retinoic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Cholecalciferol (D3), analytical standard
Supelco
Tretinoin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Cholecalciferol, analytical standard
Tretinoin, European Pharmacopoeia (EP) Reference Standard
USP
Tretinoin, United States Pharmacopeia (USP) Reference Standard