  • Search Results
  • Fluorous scavenger for parallel preparation of tertiary sulfonamides leading to secondary amines.
Fluorous scavenger for parallel preparation of tertiary sulfonamides leading to secondary amines.
Tetrahedron Letters (2007)
Basle, Emmanuel, et al.