  • Search Results
  • Carbonic anhydrase IX (CAIX) as a mediator of hypoxia-induced stress response in cancer cells.
Carbonic anhydrase IX (CAIX) as a mediator of hypoxia-induced stress response in cancer cells.
Sub-cellular biochemistry (2013-10-23)
Paul C McDonald, Shoukat Dedhar
ABSTRACT
The development of hypoxic microenvironments within many types of solid tumors imposes a significant stress on cancer cells to which they must respond appropriately in order to survive and grow. Tumor-specific, hypoxia-induced upregulation of Carbonic Anhydrase IX (CAIX) is a component of the complex response of cancer cells to the evolving low oxygen environment. Here, we discuss evidence from in vivo tumor models employing inhibition or enhancement of CAIX expression, using gene depletion or overexpression strategies, respectively, or inhibition of its catalytic activity, using CAIX-specific small molecules or antibodies, to demonstrate that CAIX is a functional mediator of tumor growth and metastasis. We also discuss the functional contribution of CAIX to several specific biological processes critical for cancer progression, including pH regulation and cell survival, adhesion, migration and invasion, the maintenance of cancer stem cell function, and the acquisition of chemo and radioresistant properties. The demonstration of CAIX as a functional mediator of cancer progression provides a biological rationale for its use as a cancer-specific, clinically relevant therapeutic target.
Sigma-Aldrich
Carbonic Anhydrase II human, recombinant, expressed in E. coli, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase II bovine, ≥90% (SDS-PAGE), recombinant, expressed in E. coli, ≥5,000 units/mg protein, buffered aqueous solution
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder