  • Search Results
  • [Light-isotope water as a therapeutic agent for acute radiation disease].
[Light-isotope water as a therapeutic agent for acute radiation disease].
Aviakosmicheskaia i ekologicheskaia meditsina = Aerospace and environmental medicine (2014-02-04)
A A Ivanov, I B Ushakov, E I Kulikova, D M Kriuchkova, Iu S Severiukhin, S V Vorozhtsova, A N Abrosimova, V A Gaevskiĭ, Iu E Siniak, A I Grigor'ev
ABSTRACT
Eighty outbred CD-I male and female mice (initial body mass of 31-34 g) and 40 male Wistar rats (initial body mass of 280-360 g) were randomly distributed into the experimental and control groups by sex and body mass. According to the results of experiments, consumption of rectified light-isotope water over 30 days immediately after irradiation by gamma-quanta 60Co at 5 Gy and 6.5 Gy can increase survivability rate significantly, accelerate hemopoiesis recovery and prevent body mass loss in irradiated animals. Therefore, light-isotope water can be considered as an effective agent for treatment of the bone marrow syndrome.
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Water, tested according to Ph. Eur.
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Supelco
Water, suitable for ion chromatography
Supelco
Water, for TOC analysis