MilliporeSigma
  • Synthesis, tumor inhibitory and antioxidant activity of new polyfunctionally 2-substituted 5,6,7,8-tetrahydronaphthalene derivatives containing pyridine, thioxopyridine and pyrazolopyridine moieties.
Synthesis, tumor inhibitory and antioxidant activity of new polyfunctionally 2-substituted 5,6,7,8-tetrahydronaphthalene derivatives containing pyridine, thioxopyridine and pyrazolopyridine moieties.
Acta poloniae pharmaceutica (2014-01-05)
Nehal A Hamdy, Manal M Anwar, Khadiga M Abu-Zied, Hanem M Awad
ABSTRACT
2-Acetyl-5,6,7,8-tetrahydronaphthalene (1) was allowed to react with different aromatic aldehydes to produce the cyanopyridones 2a and 2b, which were treated with phosphorous pentasulfide to afford the corresponding thioxopyridine derivatives 3a and 3b, respectively. The reaction of 3a and 3b with ethyl bromoacetate afforded the ester derivatives 4a and 4b, while their condensation with hydrazine hydrate gave the corresponding pyrazolopyridine derivatives 5a and 5b. The reaction of the precursor 2-amino-5,6,7,8-tetrahydronaphthalene (6) with ethoxy methylenemalonic ester led to the formation of aminomethylenemalonate derivative 7. Cyclization of 7 in boiling diphenyl ether gave the derivative - ethyl 6,7,8,9-tetrahydro-4-hydroxybenzo[g]-quinoline-3-carboxylate (8) which was hydrolyzed to produce the corresponding carboxylic acid analogue 9. Further reaction of 3a with 3-chloropropane-1,2-diol and/or iodomethane produced the corresponding nicotinonitrile derivatives 10 and 11. Hydrazinolysis of derivative 11 gave the hydrazinyl derivative 12. Moreover, chlorination of compound 2a with phosphorous oxychloride led to 2-chloro nicotinonitrile derivative 13, which was refluxed with various amines to form the corresponding derivatives 5a, 14 and 15. Treatment of the pyrazolopyridine compound 5a with formic acid and acetic anhydride afforded the corresponding formamide and acetamide analogues 16 and 17, while its reaction with DMF-DMA yielded the corresponding formimidamide derivative 18. The pyridopyrazolo[1,5-a]pyrimidine derivative 19 was obtained by cyclization of 5a with acetyl acetone. The antioxidant activity evaluation of the newly synthesized compounds showed that the pyrazolopyridine derivative 5a exhibited scavenging potency higher than that obtained by ascorbic acid. Tumor inhibitory activity screening revealed that derivatives 8 and 10 showed promising potency against the liver cancer cells (HepG-2) compared to doxorubicin as a reference drug.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, 99%
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for plant cell culture
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl