MilliporeSigma
Sorafenib in renal cell carcinoma.
Pakistan journal of pharmaceutical sciences (2014-01-01)
Ehsan Taghizadeh Davoudi, Mohamed Ibrahim bin-Noordin, Hamid Akbari Javar, Ali Kadivar, Bahare Sabeti
ABSTRACT
Cancer is among most important causes of death in recent decades. Whoever the renal cell carcinoma incidence is low but it seems it is more complicated than the other cancers in terms of pathophysiology and treatments. The purpose of this work is to provide an overview and also deeper insight to renal cell carcinoma and the steps which have been taken to reach more specific treatment and target therapy, in this type of cancer by developing most effective agents such as Sorafenib. To achieve this goal hundreds of research paper and published work has been overviewed and due to limitation of space in a paper just focus in most important points on renal cell carcinoma, treatment of RCC and clinical development of Sorafenib. The information presented this paper shows the advanced of human knowledge to provide more efficient drug in treatment of some complicated cancer such as RCC in promising much better future to fight killing disease.
Product Description
Supelco
Nicotinamide (Niacinamide), analytical standard
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®